Order Entry
Switzerland
ContactUsLinkComponent
Hydrochloric acid 5 mol/l (5 N) for biopharmaceutical production
Irritant
Corrosive
Hydrochloric acid 5 mol/l (5 N) for biopharmaceutical production
Catalog # 85400.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Hydrochloric acid 5 mol/l (5 N) for biopharmaceutical production
Catalog # 85400.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Titer:
    4.9 - 5.1 mol/l
  • Appearance:
    Passes test
  • Free bromine or chlorine:
    Passes test
  • Identification:
    Passes test
  • Colouration:
    ≤ 10 APHA
  • Evaporation residue:
    ≤ 100 ppm
  • Free chlorine:
    ≤ 1 ppm
  • Heavy metals (as Pb):
    ≤ 1 ppm
  • Ignition residue (SO₄):
    ≤ 80 ppm
  • SO₃ (Sulphite):
    ≤ 10 ppm
  • SO₄ (Sulphate):
    ≤ 5 ppm
  • As (Arsenic):
    ≤ 1.5 ppm
  • Residual metal catalysts:
    Unlikely to be present
  • Residual solvents (ICH Q3C):
    Excl.by manufact. process
  • Endotoxin:
    ≤ 0.50 EU/ml
  • Filtered through 0.2 μm:
    Confirmed

Specification Test Results

About this item

 :
Density:1.08 g/cm³ (20 °C)
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324