Order Entry
Switzerland
ContactUsLinkComponent
Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution, AVS TITRINORM volumetric solution
Corrosive
Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 5046.9010
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sulphuric acid 0.13 mol/l (0.26 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 5046.9010
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer:
    0,1235 - 0,1365 mol/l
  • Density (20/4):
    1 - 1,02

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.005 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:0
UN:3264
ADR:8,III
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589