ContactUsLinkComponent
Clostridium difficile toxoid A
Clostridium difficile toxoid A
Catalog # ENZOBMLG1450050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Clostridium difficile toxoid A
Catalog # ENZOBMLG1450050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    50 µG
  • Description:
    Clostridium difficile toxoid A, 50 µg
  • Seq:
    02
Specifications
About this item
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
  • Minimal lot-to-lot variation
  • Inactivated by formaldehyde
  • Lyophilised for ease of handling and long term stability