About this item
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci of gastric carcinoma and in distinguishing pulmonary adenocarcinomas (60-70% are CEA ) from pleural mesotheliomas (rarely or weakly CEA ). Anti-CEA positivity is seen in adenocarcinomas from the lung, colon, stomach, esophagus, pancreas, gallbadder, urachus, salivary gland, ovary, and endocervix.
Type: Primary
Antigen: CD66 molecule
Clonality: Monoclonal
Clone: C66/1030
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC471030-100
- BNC471030-500
- Antigen symbol:
- CD66 molecule
- CD66 molecule
- Antigen name:
- CD66
- CD66
- Antigen synonyms:
- CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
- CEACAM5|CD66|Biliary Glycoprotein (BGP-1)|Carcinoembryonic Antigen-related Cell Adhesion Molecule 5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone C66/1030
- Clone C66/1030
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 709196
- 709196
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide