Anti-PYCARD Rabbit Polyclonal Antibody (DyLight® 550)
N° de catalogue NOVUNBP1-78977R
Fournisseur: Novus Biologicals
Spécifications
- Antibody type:Primary
- Antigen name:PYD and CARD domain containing
- Antigen symbol:PYCARD
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:29108
- Antigen synonyms:ASC, ASC, TMS, TMS1, CARD5, TMS-1, ASC, Apoptosis-associated speck-like protein containing a CARD, Caspase recruitment domain-containing protein 5, hASC
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Spécifications
À propos de cet article
Type: Primary
Antigen: PYCARD
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse