Anti-Prostein Rabbit Polyclonal Antibody
Catalog # NOVUNB200-129
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Prostein
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal Prostein Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Type: Primary
Antigen: Prostein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human