Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNB110-94162H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:MNAT1, CDK activating kinase assembly factor and Methionine Adenosyltransferase 2A
- Antigen symbol:MNAT1/MAT2A
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4144
- Antigen synonyms:SAMS2|MATA2|MATII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal MAT1/2A Antibody [HRP]. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Bovine, Primate, Zebrafish, Mouse (-).
Type: Primary
Antigen: MNAT1/MAT2A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human