Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB110-94162C
Anbieter: Novus Biologicals
Technische Daten
- Antibody type:Primary
- Antigen name:MNAT1, CDK activating kinase assembly factor and Methionine Adenosyltransferase 2A
- Antigen symbol:MNAT1/MAT2A
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:4144
- Antigen synonyms:SAMS2|MATA2|MATII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Technische Daten
Über diesen Artikel
Rabbit Polyclonal MAT1/2A Antibody [DyLight 650]. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Bovine, Primate, Zebrafish, Mouse (-).
Type: Primary
Antigen: MNAT1/MAT2A
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human