Order Entry
Switzerland
ContactUsLinkComponent
Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]
Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-AIP Mouse Monoclonal Antibody [clone: 35-2]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-127
  • NOVUNB100-127SS
  • Antigen symbol:
  • AIP
  • AIP
  • Antigen name:
  • Aryl hydrocarbon receptor interacting protein
  • Aryl hydrocarbon receptor interacting protein
  • Antigen synonyms:
  • FKBP37|AIP|FKBP16|aryl hydrocarbon receptor-interacting protein|XAP2 |ARA9|aryl hydrocarbon receptor interacting protein |HBV-X associated protein
  • FKBP37|AIP|FKBP16|aryl hydrocarbon receptor-interacting protein|XAP2 |ARA9|aryl hydrocarbon receptor interacting protein |HBV-X associated protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 35-2
  • 35-2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 9049
  • 9049
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together