About this item
Hepatocyte Paraffin 1 or HepPar1 localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC s occur primarily in the stomach, but they are also found in many other organs. The Hepatocyte Specific Antigen may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of the Hepatocyte Specific Antigen correlates with smaller tumor size and longer patient survival. Occasionally, Hepatocyte Specific Antigen is also found in gastric carcinomas as well as in a few other non-hepatic tumors.
Type: Primary
Antigen: Hepatocyte Specific
Clonality: Monoclonal
Clone: HepPar1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Human
Specifications
- Catalog No:
- BNCB0966-500
- BNCB0966-100
- Antigen name:
- Hepatocyte Specific
- Hepatocyte Specific
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- Clone HepPar1
- Clone HepPar1
- Reactivity:
- Human, Dog
- Human, Dog
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide