Anti-IL37 Rabbit Polyclonal Antibody [clone: ]
Référence ADIPAG25A0107C100
Fournisseur: AdipoGen Life Sciences
Spécifications
- Antigen name:Interleukin 37
- Antigen symbol:IL37
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FIL1Z|IL-1H4|IL1H4|FIL1|IL-1F7|IL1RP1|IL-1H|FIL1(ZETA)|IL-1RP1|IL-37|IL1F7
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:100 µG
Spécifications
À propos de cet article
Type:
Antigen: IL37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human