Anti-NUCB2 Rabbit Polyclonal Antibody [clone: ]
Catalog # ADIPAG25A0053C100
Supplier: AdipoGen Life Sciences
Specifications
- Antigen symbol:NUCB2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type:
Antigen: NUCB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse