Order Entry
Switzerland
ContactUsLinkComponent
Anti-ANGPTL7 Rabbit Polyclonal Antibody [clone: ]
Anti-ANGPTL7 Rabbit Polyclonal Antibody [clone: ]
Catalog # ADIPAG25A0046C100
Anti-ANGPTL7 Rabbit Polyclonal Antibody [clone: ]
Catalog # ADIPAG25A0046C100

Specifications

  • Antigen symbol:
    ANGPTL7
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
  • Storage temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Type:
Antigen: ANGPTL7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human