Anti-FABP4 Rabbit Polyclonal Antibody [clone: ]
ADIPAG25A0038C100
- Antigen name:Fatty Acid Binding Protein 4
- Antigen symbol:FABP4
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ALBP|A-FABP|adipocyte-type fatty acid-binding protein|fatty acid-binding protein 4|AFABP|aP2|HEL-S-104
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:100 µG
Type:
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human