Anti-RELMA Rabbit Polyclonal Antibody [clone: ]
100 μg
Catalog # ADIPAG25A0009C100
Marketsource
Supplier: AdipoGen Life Sciences
Anti-RELMA Rabbit Polyclonal Antibody [clone: ]
Catalog # ADIPAG25A0009C100
Supplier: AdipoGen Life Sciences
Marketsource
Jump to:
Product Details & Documents
Type:
Antigen: RELMA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Specifications
- Size:100 μg
- Western blot:Yes
- Clonality:Polyclonal
- Form:Liquid
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Host:Rabbit
- Concentration:1 mg/ml
- Antigen name:Resistin Like Alpha
- Cat. No.:ADIPAG25A0009C100
- Reactivity:Rat
- Antigen synonyms:Cysteine-rich secreted protein FIZZ1|Resistin-like alpha
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Antigen symbol:RELMA
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- ELISA:Yes



