Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Anti-GITR Rat Polyclonal Antibody (Biotin) [clone: ]

Anti-GITR Rat Polyclonal Antibody (Biotin) [clone: ]

50 μg
Catalog # ADIPAG20B0043C100
Marketsource
Anti-GITR Rat Polyclonal Antibody (Biotin) [clone: ]
Catalog # ADIPAG20B0043C100
Marketsource
Anti-GITR Rat Polyclonal Antibody (Biotin) [clone: ]

50 μg

Price
SFr. 330.00
Log in for availability
1 * 100 µG
Product Details & Documents

Type:
Antigen: GITR
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Specifications
  • Size:
    50 μg
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Form:
    Liquid
  • Storage temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Host:
    Rat
  • Concentration:
    1 mg/ml
  • Antigen name:
    Tumor necrosis Factor Receptor superfamily member 18
  • Cat. No.:
    ADIPAG20B0043C100
  • Reactivity:
    Mouse
  • Antigen synonyms:
    CD357|Tumor necrosis factor receptor superfamily member 18|GITR|Glucocorticoid-induced TNFR-related protein|TNFRSF18|Activation-inducible TNFR family receptor|UNQ319|AITR|PRO364
  • Immunogen:
    Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
  • Conjugation:
    Biotin
  • Antigen symbol:
    GITR
  • Storage buffer:
    0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
  • ELISA:
    Yes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.