Specifications
- Antigen name:Caspase 1
- Antigen symbol:CASP1
- Clonality:Monoclonal
- Clone:Casper-1
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Storage buffer:In PBS containing 10% glycerol and 0,02% sodium azide.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Netrin-4 is a 628 amino acid laminin-related secreted protein that promotes neurite elongation at low concentrations but inhibits neurite extension at high concentrations. Recent evidence suggest that Netrin family members act as regulators of both neuronal and vascular growth and are implicated in controling epithelial cell branching morphogenesis in the pancreas, breast and lung. Netrin-4 inhibits proliferation of a variety of human tumor cells. Netrin-4 also induces proliferation, migration, adhesion, tube formation and survival of human lymphatic endothelial cells and seems to function as a lymphangiogenic factor contributing to tumor dissemination.
- Stable for at least 1 year after receipt when stored at -20°C.
Purified from concentrated hybridoma tissue culture supernatant.
Purity: >95% (SDS-PAGE)
Type:
Antigen: Netrin-4
Clonality: Monoclonal
Clone: Nely-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse