Anti-TRAIL-R2 Mouse Monoclonal Antibody (Biotin) [clone: HS201]
Catalog # ADIPAG20B0020C100
Supplier: AdipoGen Life Sciences
Specifications
- Antigen name:TNF receptor superfamily member 10B
- Antigen symbol:TNFRSF10B
- Clonality:Monoclonal
- Clone:HS201
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:TRAILR2|TRAIL-R2|CD262|Trail receptor 2|TRICK2|KILLER|TRICK2A|TRICKB|ZTNFR9|KILLER/DR5|TRICK2B|DR5
- Storage buffer:In PBS containing 0,02% sodium azide.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:0,5 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type:
Antigen: TRAIL-R2
Clonality: Monoclonal
Clone: HS201
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human