Anti-TRAIL-R1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: HS101]
Catalog # ADIPAG20B0020BC100
Supplier: AdipoGen Life Sciences
Specifications
- Antigen name:TNF receptor superfamily member 10a
- Antigen symbol:TRAIL-R1
- Clonality:Monoclonal
- Clone:HS101
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:TRAILR1|DR4|APO2|CD261|TRAILR-1|MGC9365
- Storage buffer:In PBS containing 0,02% sodium azide.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 tests
- Pk:100 µG
Specifications
About this item
Type:
Antigen: TRAIL-R1
Clonality: Monoclonal
Clone: HS101
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human