Anti-BAFF-R Mouse Monoclonal Antibody [clone: HuBR9.1]
ADIPAG20B0012FC100
- Antigen symbol:BAFF-R
- Clonality:Monoclonal
- Clone:HuBR9.1
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Storage buffer:In PBS containing 10% glycerol and 0,02% sodium azide.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:100 µG
Type:
Antigen: BAFF-R
Clonality: Monoclonal
Clone: HuBR9.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human