Order Entry
Switzerland
ContactUsLinkComponent
Anti-CSF3 Mouse Monoclonal Antibody (CF488A) [clone: CSF3/900]
Anti-CSF3 Mouse Monoclonal Antibody (CF488A) [clone: CSF3/900]
Supplier:  Biotium
Certificates
Anti-CSF3 Mouse Monoclonal Antibody (CF488A) [clone: CSF3/900]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BTIUBNC880900-500
  • BTIUBNC880900-100
  • Antigen symbol:
  • CSF3
  • CSF3
  • Antigen name:
  • Granulocyte-Colony Stimulating Factor
  • Granulocyte-Colony Stimulating Factor
  • Antigen synonyms:
  • G-CSF|colony stimulating factor 3 (granulocyte)|Filgrastim|chromosome 17 open reading frame 33 (C17orf33)|CSF3OS|Lenograstim|Pluripoietin
  • G-CSF|colony stimulating factor 3 (granulocyte)|Filgrastim|chromosome 17 open reading frame 33 (C17orf33)|CSF3OS|Lenograstim|Pluripoietin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone CSF3/900
  • Clone CSF3/900
  • Reactivity:
  • Macaque, Human
  • Macaque, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2233
  • 2233
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together