Order Entry
Switzerland
ContactUsLinkComponent
Anti-JAG1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: J1G53-3]
Anti-JAG1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: J1G53-3]
Catalog # ADIPAG20A0048C100
Anti-JAG1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: J1G53-3]
Catalog # ADIPAG20A0048C100

Specifications

  • Antigen name:
    Jagged 1
  • Antigen symbol:
    JAG1
  • Clonality:
    Monoclonal
  • Clone:
    J1G53-3
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    Ser-1|Htu|AWS|AHD|Jag1|jagged|JAGL1|ABE2|HJ1|Ozz|AGS|Gsfabe2|CD339
  • Storage buffer:
    0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
  • Storage temperature:
    Short term: +4 °C, Long Term: –20 °C
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
  • Size:
    50 μg
  • Pk:
    100 µG

Specifications

About this item

Type:
Antigen: JAG1
Clonality: Monoclonal
Clone: J1G53-3
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human