Anti-TIM3 Rat Monoclonal Antibody [clone: TI 339H]
Catalog # ADIPAG20A0029C050
Supplier: AdipoGen Life Sciences
Specifications
- Antigen symbol:TIM3
- Clonality:Monoclonal
- Clone:TI 339H
- ELISA:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Storage buffer:0,2µm-filtered solution in PBS, pH 7,4. , contains no preservatives.
- Storage temperature:Short term: +4 °C, Long Term: –20 °C
- Concentration:1 mg/ml
- Immunogen:Recombinant human adiponectin. Recognizes human adiponectin. Detects a band of ~30kDa by Western blot. Cross-reactivity with other species not tested.
- Size:100 μg
- Pk:50 µG
Specifications
About this item
Type:
Antigen: TIM3
Clonality: Monoclonal
Clone: TI 339H
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse