About this item
This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40 kDa. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: A53-B/A2.26 BA17
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- BNC400753-500
- BNC400753-100
- Antigen symbol:
- KRT19
- KRT19
- Antigen name:
- Cytokeratin 19
- Cytokeratin 19
- Antigen synonyms:
- k19|k1cs|krt19|Keratin 19 Keratin Type i 40kD
- k19|k1cs|krt19|Keratin 19 Keratin Type i 40kD
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone A53-B/A2.26 BA17
- Clone A53-B/A2.26 BA17
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 654568
- 654568
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide