Technische Daten
- Antibody type:Primary
- Antigen name:SET-binding protein 1
- Antigen symbol:SETBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:26040
- Antigen synonyms:MRD29|SETBP1|SET-binding protein|KIAA0437|SEB
- Storage buffer:Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Technische Daten
Über diesen Artikel
Midline-2 is a 715 amino acid protein encoded by the human gene MID2. Midline-2 belongs to the TRIM/RBCC family and contains two B box-type zinc fingers, one B30.2/SPRY domain, one COS domain, one fibronectin type-III domain and one RING-type zinc finger. Midline-2 is a cytoplasmic protein found as a homodimer or heterodimer with Midline-1. It also interacts with IGBP1 (Lymphocyte signaling protein A4). Dimerization is mediated by the tripartite motif (RBCC; RING- and B box-type zinc fingers and coiled coil domains) and microtubule association is dependent on the C-terminal B30.2 domain. Midline-2 is expressed at low levels in fetal kidney and lung, and in adult prostate, ovary and small intestine.
Recommended Dilutions: Western Blot: 1:100 to 1000; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.
Type: Primary
Antigen: SETBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat