Anti-CSF2 Mouse Monoclonal Antibody (CF647) [clone: BVD2-21C11]
: Biotium
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: Clone BVD2-21C11
Conjugation: CF647
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey
- Catalog No:
- BNC470662-100
- BNC470662-500
- Antigen symbol:
- CSF2
- CSF2
- Antigen name:
- GM-CSF
- GM-CSF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone BVD2-21C11
- Clone BVD2-21C11
- Reactivity:
- Cynomolgus monkey, Rhesus monkey, Human
- Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1349
- 1349
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Frequently Bought Together