Anti-PD1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-1867R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Programmed cell death protein 1
- Antigen symbol:PD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:5133
- Antigen synonyms:PD-1|hPD-1|hPD-l|programmed cell death protein 1|CD279|protein PD-1|PD1|SLEB2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:240-288/288
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat