Order Entry
Switzerland
ContactUsLinkComponent
Anti-EGFR Mouse Monoclonal Antibody (CF488A) [clone: H9B4]
Anti-EGFR Mouse Monoclonal Antibody (CF488A) [clone: H9B4]
Supplier:  Biotium
Certificates
Anti-EGFR Mouse Monoclonal Antibody (CF488A) [clone: H9B4]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880642-100
  • BNC880642-500
  • Antigen symbol:
  • EGFR
  • EGFR
  • Antigen synonyms:
  • Errp|Urogastrone|PIG61|HER1|Wa5|mENA|Receptor Tyrosine Protein Kinase|ErbB1|Erbb1|wa2|ERBB1|Proto-oncogene c-ErbB-1
  • Errp|Urogastrone|PIG61|HER1|Wa5|mENA|Receptor Tyrosine Protein Kinase|ErbB1|Erbb1|wa2|ERBB1|Proto-oncogene c-ErbB-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone H9B4
  • Clone H9B4
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 488293
  • 488293
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together