Anti-CXCL8 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0780R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) ligand 8
- Antigen symbol:CXCL8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:3576
- Antigen synonyms:MDNCF|LYNAP|IL8|NAP1|NAP-1|GCP1|MONAP|LECT|NAF|GCP-1|LUCT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:30-60/99
- Purification:Purified by Protein A
- Size:100 μL
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CXCL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit