Anti-ESR1 Mouse Monoclonal Antibody (CF405S) [clone: ER505]
Supplier: Biotium
Certificates
About this item
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER505
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BTIUBNC040505-500
- BTIUBNC040505-100
- Antigen symbol:
- ESR1
- ESR1
- Antigen name:
- Estrogen Receptor
- Estrogen Receptor
- Antigen synonyms:
- Nuclear receptor subfamily 3 group A member 1|Estrogen Receptor alpha delta 4*5,6,7*/654 isoform|Estrogen Receptor alpha delta 4 49 isoform
- Nuclear receptor subfamily 3 group A member 1|Estrogen Receptor alpha delta 4*5,6,7*/654 isoform|Estrogen Receptor alpha delta 4 49 isoform
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone ER505
- Clone ER505
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 208124
- 208124
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together