IgG Isotype Control (Unconjugated)
Catalog # GNTXGTX76672
Supplier: Genetex
Specifications
- Antibody type:Control
- Conjugation:Unconjugated
- Isotype:IgG
- Storage buffer:Reconstitute to a minimum of 1 mg/ml with a saline (150 mM NaCl) solution.
- Storage temperature:Keep as concentrated solution. Prior to reconstitution, store at 4 °C short term. For extended storage after reconstitution, aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Purification:Sera
- Size:10 mg
- Pk:10 mg
Specifications
About this item
Type: Control
Antigen:
Clonality:
Clone: Unconjugated
Conjugation:
Epitope:
Host:
Isotype: IgG
Reactivity: