Order Entry
Switzerland
ContactUsLinkComponent
Anti-A8 + S100A Mouse Monoclonal Antibody [clone: 27E10]
Anti-A8 + S100A Mouse Monoclonal Antibody [clone: 27E10]
  GNTXGTX17050
 :  Genetex
Anti-A8 + S100A Mouse Monoclonal Antibody [clone: 27E10]
  GNTXGTX17050
 :  Genetex

 

  • Antibody type:
    Primary
  • Antigen name:
    S100A8 + S100A9
  • Antigen symbol:
    A8 + S100A
  • Clonality:
    Monoclonal
  • Clone:
    27E10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Monkey,
    Human
  • Storage buffer:
    Preservative: 0,02% Sodium Azide. Constituents: 0,1% BSA, PBS
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4 °C.
  • Concentration:
    0,1 mg/ml
  • Immunogen:
    Unfortunately, this information is considered to be commercially sensitive
  • Purification:
    Protein G purified
  • Size:
    50 μg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: A8 + S100A
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 27E10
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey