Anti-ATR Rabbit Polyclonal Antibody
GNTXGTX128145
: Genetex
- Antibody type:Primary
- Antigen name:Atr Serine/Threonine Kinase
- Antigen symbol:ATR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:545
- Antigen synonyms:FRP1|FCTCS|SCKL|MEC1|SCKL1
- Storage buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0,025% ProClin 300 was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0,63 mg/ml
- Purification:Purified by antigen-affinity chromatography.
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: ATR
Clonality: Polyclonal
Clone: Unconjugated
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse