Anti-HAX1A Mouse Monoclonal Antibody [clone: 9G3D11]
Katalognr. GNTXGTX84983
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:HCLS1 associated protein X-1
- Antigen symbol:HAX1
- Clonality:Monoclonal
- Clone:9G3D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Gene ID:10456
- Antigen synonyms:SCN3|HS1BP1|HCLSBP1
- Storage buffer:Antibody is supplied in PBS containing 0,02% sodium azide
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Immunogen:Mouse monoclonal Hax1a antibody was raised against a 15 amino acid peptide near the amino terminus of human Hax1a.
- Purification:Immunoaffinity chromotography purified IgG
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HAX1A
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 9G3D11
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat