Anti-ACAT2 Mouse Monoclonal Antibody [clone: 1C5]
GNTXGTX84977
: Genetex
- Antibody type:Primary
- Antigen name:Acetyl-CoA acetyltransferase 2
- Antigen symbol:ACAT2
- Clonality:Monoclonal
- Clone:1C5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Monkey,Human,Dog,Rat
- Western blot:Yes
- Gene ID:39
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human ACAT2 (NP_0058826) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: ACAT2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1C5
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Dog, Monkey, Rat