Anti-AK1 Mouse Monoclonal Antibody [clone: 18C7]
GNTXGTX84937
: Genetex
- Antibody type:Primary
- Antigen name:Adenylate Kinase 1
- Antigen symbol:AK1
- Clonality:Monoclonal
- Clone:18C7
- Conjugation:Unconjugated
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:203
- Antigen synonyms:HTL-S-58j
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Protein expressed in 293T cell transfected with human AK1 expression vector
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: AK1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 18C7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human