Anti-BCKDK Mouse Monoclonal Antibody [clone: 11C9]
Catalog # GNTXGTX84836
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Branched-chain alpha-ketoacid dehydrogenase kinase
- Antigen symbol:BCKDK
- Clonality:Monoclonal
- Clone:11C9
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:10295
- Antigen synonyms:3-methyl-2-oxobutanoate dehydrogenase lipoamide kinase|BCKD-kinase|BCKDHKIN|mitochondrial|Branched-chain alpha-ketoacid dehydrogenase kinase
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Protein expressed in 293T cell transfected with human BCKDK expression vector
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: BCKDK
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 11C9
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human