Anti-BTN1A1 Mouse Monoclonal Antibody [clone: 1A6]
Codice catalogo GNTXGTX84779
Nome del fornitore: Genetex
Specifiche
- Antibody type:Primary
- Antigen name:butyrophilin, subfamily 1, member A1
- Antigen symbol:BTN1A1
- Clonality:Monoclonal
- Clone:1A6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:696
- Antigen synonyms:BT|BTN|BTN1|Btn
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human BTN1A1 (NP_001723) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: BTN1A1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1A6
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human