Anti-CLPP (ClpP Caseinolytic peptidase ATP-dependent proteolytic Subunit homolog (E. coli)) Mouse Monoclonal Antibody [clone: 1E8]
Katalognr. GNTXGTX84680
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Caseinolytic Mitochondrial Matrix Peptidase Proteolytic Subunit
- Antigen symbol:CLPP (ClpP Caseinolytic peptidase ATP-dependent proteolytic Subunit homolog (E. coli))
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western blot:Yes
- Gene ID:8192
- Antigen synonyms:PRLTS3|DFNB81
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human CLPP (NP_006003) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CLPP (ClpP Caseinolytic peptidase ATP-dependent proteolytic Subunit homolog (E. coli))
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1E8
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey