Anti-CNDP1 (carnosine dipeptidase 1 (metallopeptidase M20 Family)) Mouse Monoclonal Antibody [clone: 2F8]
Katalognr. GNTXGTX84677
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Carnosine Dipeptidase 1 (Metallopeptidase M20 Family)
- Antigen symbol:CNDP1 (carnosine dipeptidase 1 (metallopeptidase M20 Family))
- Clonality:Monoclonal
- Clone:2F8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:84735
- Antigen synonyms:CN1|CPGL2|HsT2308
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Protein expressed in 293T cell transfected with human CNDP1 expression vector
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CNDP1 (carnosine dipeptidase 1 (metallopeptidase M20 Family))
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 2F8
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human