Anti-EPM2AIP1 (EPM2A (laforin) interacting Protein 1) Mouse Monoclonal Antibody [clone: 2A5]
GNTXGTX84564
: Genetex
- Antibody type:Primary
- Antigen name:EPM2A (laforin) interacting protein 1
- Antigen symbol:EPM2AIP1
- Clonality:Monoclonal
- Clone:2A5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:9852
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human EPM2AIP1 (NP_055620) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: EPM2AIP1 (EPM2A (laforin) interacting Protein 1)
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 2A5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human