Anti-FOXA1 Mouse Monoclonal Antibody [clone: 3A8]
Catalog # GNTXGTX84487
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box A1
- Antigen symbol:FOXA1
- Clonality:Monoclonal
- Clone:3A8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human,Rat
- Western blot:Yes
- Gene ID:3169
- Antigen synonyms:HNF-3a|MGC33105|FoxA4a|MGC52590|pintallavis|tcf3a|fkh|xfkh2|FOXA1|Tcf3a|Tcf-3a|fkd7|forkhead-7|tFoxA1|HNF3A|HNF3-alpha|TCF3A|MGC75967|Ci-fkh|Hnf-3a|xfkh1|foxa1|HNF3alpha|Hnf3a
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Recombinant protein expressed in E.coli corresponding to amino acids 1-473 of human FOXA1
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: FOXA1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 3A8
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat