Anti-GAD1 Mouse Monoclonal Antibody [clone: 3H2]
Katalognr. GNTXGTX84470
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Glutamate decarboxylase 1
- Antigen symbol:GAD1
- Clonality:Monoclonal
- Clone:3H2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Gene ID:2571
- Antigen synonyms:67kDa)|SCP|glutamate decarboxylase 1|67kD)|glutamate decarboxylase 1 (brain|CPSQ1|GAD|GAD1|OTTHUMP00000041055|GAD67|GAD25|FLJ45882
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Protein expressed in 293T cell transfected with human GAD1 expression vector
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GAD1
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 3H2
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human