Order Entry
Switzerland
ContactUsLinkComponent
Anti-PODXL Mouse Monoclonal Antibody (Biotin) [clone: 3D3]
Anti-PODXL Mouse Monoclonal Antibody (Biotin) [clone: 3D3]
Supplier:  Biotium
Certificates
Anti-PODXL Mouse Monoclonal Antibody (Biotin) [clone: 3D3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0427-500
  • BNCB0427-100
  • Antigen symbol:
  • PODX
  • PODX
  • Antigen name:
  • Podocalyxin
  • Podocalyxin
  • Antigen synonyms:
  • PODXL|PDXL|PCLP1|Gp200|GCTM-2|Podocalyxin-like protein 1|Pcx
  • PODXL|PDXL|PCLP1|Gp200|GCTM-2|Podocalyxin-like protein 1|Pcx
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone 3D3
  • Clone 3D3
  • Reactivity:
  • Rabbit, Human, Rat
  • Rabbit, Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 732423
  • 732423
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together