Anti-KLHL2 Mouse Monoclonal Antibody [clone: 1G7]
Catalog # GNTXGTX84259
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Kelch-like 2
- Antigen symbol:KLHL2
- Clonality:Monoclonal
- Clone:1G7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Monkey,Human,Dog
- Western blot:Yes
- Gene ID:11275
- Antigen synonyms:MAV|ABP-KELCH|MAYVEN
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1-100 and 494-593 of human KLHL2 (NP_009177) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: KLHL2
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1G7
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Dog, Monkey