Order Entry
Switzerland
ContactUsLinkComponent
Anti-TGM2 Mouse Monoclonal Antibody (CF488A) [clone: TGM2/419]
Anti-TGM2 Mouse Monoclonal Antibody (CF488A) [clone: TGM2/419]
Supplier:  Biotium
Certificates
Anti-TGM2 Mouse Monoclonal Antibody (CF488A) [clone: TGM2/419]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880419-500
  • BNC880419-100
  • Antigen symbol:
  • TGM2
  • TGM2
  • Antigen name:
  • Transglutaminase II
  • Transglutaminase II
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone TGM2/419
  • Clone TGM2/419
  • Reactivity:
  • Rabbit, Monkey, Human, Rat, Mouse
  • Rabbit, Monkey, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 517033
  • 517033
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together