Anti-MAPK12 Mouse Monoclonal Antibody [clone: 1E2]
Catalog # GNTXGTX84148
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 12
- Antigen symbol:MAPK12
- Clonality:Monoclonal
- Clone:1E2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:6300
- Antigen synonyms:P38GAMMA|PRKM12|MAPK 12|ERK3|SAPK-3|ERK6|SAPK3|ERK-6
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Protein expressed in 293T cell transfected with human MAPK12 expression vector
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MAPK12
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1E2
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human