Anti-MOBKL1A Mouse Monoclonal Antibody [clone: 1H2]
Catalog # GNTXGTX84105
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:MOB1 Mps One Binder kinase activator-like 1A (yeast)
- Antigen symbol:MOBKL1A
- Clonality:Monoclonal
- Clone:1H2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Gene ID:92597
- Antigen synonyms:MOBKL1A|MATS2|MOB4A
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human MOBKL1A(NP_775739) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MOBKL1A
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1H2
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human