Anti-MOBKL2B Mouse Monoclonal Antibody [clone: 6E3]
GNTXGTX84100
: Genetex
- Antibody type:Primary
- Antigen name:MOB1 Mps One Binder kinase activator-like 2B (yeast)
- Antigen symbol:MOBKL2B
- Clonality:Monoclonal
- Clone:6E3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:79817
- Antigen synonyms:C9orf35|MOBKL2B|MOB1D
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human MOBKL2B (NP_079037) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: MOBKL2B
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 6E3
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human