Anti-c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 1A6]
Catalog # GNTXGTX84066
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:V-Myc Avian Myelocytomatosis Viral Oncogene Homolog
- Antigen symbol:c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
- Clonality:Monoclonal
- Clone:1A6
- Conjugation:Unconjugated
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Gene ID:4609
- Antigen synonyms:MRTL|bHLHe39|c-Myc|MYCC
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Recombinant fragment expressed in E.coli corresponding to amino acids 1-454 of human c-Myc
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 1A6
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human